Order Entry
United States
ContactUsLinkComponent
ProCycle™ Full Skirted PCR Plates, Full Edge, Clear, 96×0.1 ml Well, Printed Alphanumerics
ProCycle™ Full Skirted PCR Plates, Full Edge, Clear, 96×0.1 ml Well, Printed Alphanumerics
Catalog # 104027-254
Supplier:  Alkali Scientific
undefined
ProCycle™ Full Skirted PCR Plates, Full Edge, Clear, 96×0.1 ml Well, Printed Alphanumerics
Catalog # 104027-254
Supplier:  Alkali Scientific
Supplier Number:  PCFS-96

Specifications

  • No. of wells:
    96
  • Description:
    Full skirted PCR plates, full edge, 96×0.2 ml well, printed alphanumerics, 10 per pack
  • Cat. No.:
    104027-254

Specifications

About this item

Facilitate PCR experiments with Alkali Scientific™ ProCycle™ full skirted PCR plates, featuring full edge design, clear material, 96×0.1 ml well, and printed alphanumerics, available in packs of 10.

  • Universal compatibility: PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your laboratory
  • Easy well identification: High-contrast, black alphanumeric inde×ing facilitates quick and accurate well identification, strea mlining workflow
  • Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
  • Sample protection: Well ridges help reduce sample evaporation when sealing with film, foil or strip caps, preserving the integrity of your samples
  • High clarity, ultra-pure material: PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
  • Purity assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free

Experience unparalleled performance and versatility in your PCR applications with the ProCycle™ full skirted PCR plates. Designed with a full edge and clear construction, these plates offer exceptional durability and visibility, ensuring reliable results with each use. Featuring 96 wells with a 0.1 ml capacity, these plates provide ample space for high-throughput sample processing while minimizing sample volume. The printed alphanumerics enhance sample tracking and plate orientation, facilitating sea mless integration into experimental workflows. Engineered for even and rapid thermal transfer, these plates ensure uniform heating across all wells, optimizing PCR amplification efficiency and reducing experimental variability. Whether used for standard PCR assays or complex molecular biology applications, the ProCycle™ full skirted PCR plates are the ideal choice for researchers seeking superior performance and convenience in their laboratory work.

Compatible with the following machines:
• ABI 7000, ABI 7300, ABI 7500, ABI 7700 and ABI StepOne
• Bio-Rad iCycler, Bio-Rad myIQ and Bio-Rad myIQ5
• ABI Sequencer 3100 and ABI Sequencer 3130