Specifications
- No. of wells:96
- Description:Standard PCR plates, non-skirted, 96×0.2 ml well, black printed alphanumerics, A12 cut corner, 10 per pack
- Cat. No.:104027-828
Specifications
About this item
Facilitate PCR experiments with Alkali Scientific™ ProCycle™ standard PCR plates, non-skirted, featuring 96×0.2 ml well and black printed alphanumerics, with an A12 notch.
- Universal compatibility: PCR plate is compatible with a broad range of thermal cyclers, ensuring versatility and adaptability in your laboratory
- Easy well identification: High-contrast, black alphanumeric indexing facilitates quick and accurate well identification, strea mlining workflow
- Optimal heat transfer: Ultra-thin, uniform wells ensure optimal heat transfer, promoting high reaction efficiency and reproducibility
- Sample Protection: Well ridges help reduce sample evaporation when sealing with film, foil, or strip caps, preserving the integrity of your samples
- High clarity, ultra-pure material: This PCR plate is manufactured from high-clarity, ultra-pure polypropylene, meeting FDA 21 CFR 177.1520 and USP Class VI standards
- Purity assurance: The plate is certified free of detectable DNase, RNase, DNA, PCR inhibitors and has been tested to be pyrogen-free
Elevate your PCR experiments with the ProCycle™ standard PCR plates, meticulously designed to meet the demands of modern molecular biology research. Featuring a non-skirted design and 96 wells of 0.2 ml capacity each, these plates offer ample space for high-throughput sample processing. The black printed alphanumerics and A12 notch ensure effortless sample identification and plate orientation, streamlining experimental workflows. Engineered for optimal thermal conductivity, these plates facilitate rapid and uniform heat transfer during PCR amplification, ensuring reliable and reproducible results. Whether used for routine PCR assays or complex experimental protocols, the ProCycle™ standard PCR plates are the perfect choice for researchers seeking performance, convenience and consistency in their laboratory work.
Compatible with the following machines:
• MJ/Bio-Rad Opticon, Opticon 2, MiniOpticon and Chromo4
• Agilent/Stratagene MX4000P, MX3000P, MX3000i and MX3005P