ContactUsLinkComponent
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Flat Caps
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Flat Caps
  104027-832
undefined
ProCycle™ 0.2 ml Clear 8-Strip Tubes with Attached Hinged Flat Caps
  104027-832
 :  PC7067-F
  • Color:
    Clear
  • Description:
    PCR tubes, individual 0.2 ml flat cap
  • Cat. No.:
    104027-832
  • Packaging:
    125×8, 1000 per pack
Ensure secure sealing and easy access with Alkali Scientific™ ProCycle™ 0.2 ml clear 8-Strip tubes featuring attached hinged flat caps, available in packs of 1000.
  • 0.2 ml 8-Strip PCR tubes with attached flat caps
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Certified RNAse, DNAse and pyrogen free
Transform PCR workflows with the ProCycle™ 0.2 ml clear 8-Strip PCR tubes and attached hinged flat caps, engineered for rapid thermal transfer and precise sample containment. With each pack containing 1000 tubes, researchers can embark on their scientific endeavors with confidence, knowing that sample security and convenience are always assured. The hinged flat caps provide easy access to samples while ensuring reliable sealing, making them ideal for high-throughput experiments.