ContactUsLinkComponent
ProCycle™ 0.1 ml Low-Profile Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
ProCycle™ 0.1 ml Low-Profile Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Catalog # 104027-944
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.1 ml Low-Profile Rigid Stable 8-Strip PCR Tubes with Attached Individual Flat Caps
Catalog # 104027-944
Supplier:  Alkali Scientific
Supplier Number:  PC7065
Specifications
  • Color:
    Clear
  • Description:
    8-Strip PCR tubes, low profile with attached flat caps, 0.1 ml
  • Cat. No.:
    104027-944
  • Packaging:
    120×8, 960 per pack
Specifications
About this item
Experience stability in PCR reactions with Alkali Scientific™ ProCycle™ 0.1 ml low-profile rigid stable 8-Strip PCR tubes with attached individual flat caps.
  • Robust inter-well linkages make the strips rigid and easy to handle, preventing spills
  • Angularly-attached caps prevent lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack
  • Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free
Experience unmatched stability and performance with the ProCycle™ 0.1 ml low-profile rigid stable 8-Strip PCR tubes. Featuring attached individual flat caps for enhanced sample security, these tubes are engineered for rapid thermal transfer and optimal PCR efficiency. With their ergonomic design and superior thermal conductivity, researchers can expedite their research workflows with confidence, knowing that reliable results are always within reach.