- Color:Clear
- Description:SnapStrip™ 8-Strip tubes with single easy cut connectors and individually attached flat caps
- Cat. No.:104027-684
- Packaging:120×8, 960 per pack
Streamline PCR workflow with Alkali Scientific™ ProCycle™ SnapStrip 8-Strip tubes featuring single EZCut™ connectors and individually attached flat caps, providing clear visibility of samples.
- 0.2 ml 8-Strip PCR tubes with attached caps (120 strip × 8, 960/pk)
- Caps are optically clear for qPCR
- Ultra-thin walls for uniformity and consistency
- Positive sealing eliminates any evaporation
- Fits standard spacing on 8-channel pipettes
- Optical flat caps
- Certified RNAse, DNAse and pyrogen free
Experience enhanced convenience and efficiency in your PCR workflow with Alkali Scientific™ ProCycle™ SnapStrip 8-Strip tubes. These tubes feature single EZCut™ connectors that allow for easy separation into individual tubes, eliminating the need for additional tools and minimizing the risk of sample cross-contamination. Each tube is equipped with an individually attached flat cap, providing a secure seal to prevent sample evaporation and contamination during thermal cycling. The clear design of the tubes enables easy visualization of your samples, ensuring accurate pipetting and tracking of reactions.