ContactUsLinkComponent
ProCycle™ 0.2 ml SnapStrip™ 8-Strip PCR Tubes with Attached Individual Dome Caps
ProCycle™ 0.2 ml SnapStrip™ 8-Strip PCR Tubes with Attached Individual Dome Caps
Catalog # 104028-094
Supplier:  Alkali Scientific
undefined
ProCycle™ 0.2 ml SnapStrip™ 8-Strip PCR Tubes with Attached Individual Dome Caps
Catalog # 104028-094
Supplier:  Alkali Scientific
Supplier Number:  PC7060
Specifications
  • Color:
    Clear
  • Description:
    StableStrip™ 8-Strip PCR tubes with attached individual dome caps, 0.2 ml
  • Cat. No.:
    104028-094
  • Packaging:
    120×8, 960 per pack
Specifications
About this item
Conduct efficient PCR experiments with Alkali Scientific™ ProCycle™ 0.2 ml SnapStrip™ 8-Strip PCR tubes with attached individual dome caps, featuring 120× 8-Strips for convenient use.
  • 0.2 ml 8-Strip PCR tubes with attached caps
  • Caps are optically clear for qPCR
  • Ultra-thin walls for uniformity and consistency
  • Positive sealing eliminates any evaporation
  • Fits standard spacing on 8-channel pipettes
  • Optical dome caps
  • Certified RNAse, DNAse and pyrogen free
Enhance the efficiency of your PCR workflows with Alkali Scientific™ ProCycle™ 0.2 ml SnapStrip™ 8-Strip PCR tubes. These tubes feature a unique SnapStrip™ design with attached individual dome caps, allowing quick and easy tube opening and closure. With 120 strips per pack, each containing 8 tubes, this bulk packaging provides an ample supply for high-throughput PCR applications. The rigid and stable construction of these tubes ensures uniform PCR amplification and reliable results across all samples. Trust Alkali Scientific™ ProCycle™ SnapStrip™ PCR tubes for exceptional performance and convenience in your molecular biology experiments.