Order Entry
United States
ContactUsLinkComponent
96-Well Deep Well Plates, 1.6 ml, U-Bottom, Square Well
96-Well Deep Well Plates, 1.6 ml, U-Bottom, Square Well
Catalog # 104026-918
Supplier:  Alkali Scientific
undefined
96-Well Deep Well Plates, 1.6 ml, U-Bottom, Square Well
Catalog # 104026-918
Supplier:  Alkali Scientific
Supplier Number:  MPD43

Specifications

  • Description:
    96-well deep well plates, 1.6 ml
  • Material:
    PP
  • Cat. No.:
    104026-918
  • Packaging:
    5 per pack, 50 per case
  • Well shape:
    Square
  • Bottom style:
    U-bottom

Specifications

About this item

Optimize laboratory workflows with the 1.6 ml 96-well deep well plates by Alkali Scientific, featuring a U-Bottom and Square well design, available in packs of 5 and cases of 50.

  • Virgin, heat-resistant, medical-grade polypropylene plates
  • RNase, DNase and pyrogen free
  • Minimal binding and liquid retention
  • Black printed alpha-numeric reference grids
  • Autoclavable
  • Temperature range: –86 to 82.2 °C

Experience unmatched performance and reliability with the Alkali Scientific™ 1.6 ml 96-Well Deep Well Plate. Engineered with precision and crafted from premium-quality materials, these plates are designed to meet the demanding requirements of modern research applications. The U-bottom and square well configuration ensures efficient liquid handling and optimal sample recovery, resulting in precise and reproducible experimental outcomes. Rely on the exceptional quality and sterility assurance of these deep well plates to drive the success of your scientific experiments.

Packaging: Each pack contains 5 plates, with 50 packs per case, providing a consistent and dependable solution for your laboratory workflows.