Order Entry
United States
ContactUsLinkComponent
96-Well Deep Well Plates, V Conical Bottom, Square Well
96-Well Deep Well Plates, V Conical Bottom, Square Well
Catalog # 104028-984
Supplier:  Alkali Scientific
undefined
96-Well Deep Well Plates, V Conical Bottom, Square Well
Catalog # 104028-984
Supplier:  Alkali Scientific
Supplier Number:  MPD29

Specifications

  • Sterility:
    Sterile
  • Description:
    96-well deep well plates, 2.2 ml
  • Material:
    PP
  • Cat. No.:
    104028-984
  • Packaging:
    50 per case
  • Well shape:
    Square
  • Bottom style:
    V conical bottom

Specifications

About this item

Facilitate efficient sample processing with Alkali Scientific 2.2 ml 96-well deep well plates, featuring a V conical bottom and square well design, available in a case of 50.

  • Virgin, heat-resistant, medical-grade polypropylene plates
  • RNase, DNase and pyrogen free
  • Minimal binding and liquid retention
  • Black printed alpha-numeric reference grids
  • Autoclavable
  • Temperature range: –86 to 82.2 °C

Unleash the full potential of your laboratory experiments with the Alkali Scientific 2.2 ml 96-Well Deep Well Plate, featuring a V-conical bottom and square well design. Engineered to the highest standards of quality and precision, these plates are tailored to meet the demanding requirements of modern research applications. The unique V-conical bottom design enhances sample recovery and ensures minimal dead volume, optimizing liquid handling processes for maximum efficiency. With 50 plates per case, these deep well plates provide a reliable solution for a wide range of laboratory tasks, from sample storage to high-throughput screening. Experience consistent and reproducible results with these versatile plates, designed to elevate your scientific endeavors with confidence and precision.