Human Recombinant ZAK (GST tag), Active (from Baculovirus-Insect Cells)
New Product
Recombinant full length human ZAK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together