Human Recombinant 14-3-3 sigma (from E. coli)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant full length human tag-free 14-3-3 sigma was expressed in E. coli.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:YWHAS
- Protein/Peptide Name:SFN
- Molecular Weight:~29 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5 - 8.0, 50 - 300 mM NaCl, 0.25 - 1.0 mM DTT, 10 - 25% glycerol.
Specifications
Frequently Bought Together