Order Entry
United States
ContactUsLinkComponent
Human Recombinant Tau-441, GSK3beta-phosphorylated (from E. coli)
Human Recombinant Tau-441, GSK3beta-phosphorylated (from E. coli)
Supplier:  Sino Biological
New Product
Certificates
Human Recombinant Tau-441, GSK3beta-phosphorylated (from E. coli)
Supplier:  Sino Biological
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
  • Protein Synonyms:
    PPP1R103|FTDP-17|MAPT|TAU|PPND|MAPTL|MTBT1|MTBT2|MSTD|DDPAC
  • Protein/Peptide Name:
    MAPT
  • Molecular Weight:
    ~64-68 kDa
  • Formulation:
    Supplied as sterile 50 mM Tris-HCl pH 7.5, 50 - 300 mM NaCl, 1 mM DTT, 10 - 25% glycerol

Specifications

Related Information

Frequently Bought Together