Human Recombinant CDK11A (CDC2L2) (GST tag), Unactive (from Baculovirus-Insect Cells)
Supplier: Sino Biological
New Product
Certificates
About this item
Recombinant human CDK11A (361-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
- Shipping temperature: Dry ice
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Baculovirus (Insect cells)
- Species:Human
- Storage Conditions:Store product at –70 °C. For optimal storage, aliquot target into smaller quantities after centrifugation and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
- Protein Synonyms:CDC2L2|CDK11-p46|RP1-283E3.2|CDC2L3|PITSLRE|p58GTA|CDK11-p58|CDK11-p110
- Protein/Peptide Name:CDK11A
- Molecular Weight:73 kDa
- Formulation:Supplied as sterile 50 mM Tris-HCl, pH 7.5, 50 - 300 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 - 1.0 mM DTT, 0 - 0.1 mM PMSF, 10 - 25% glycerol
Specifications
Related Information
Frequently Bought Together