HiTrap™ Capto™ Octyl Hydrophobic Interaction Chromatography Columns
Supplier: Cytiva
Certificates
About this item
Capto™ Octyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
- Improved productivity and process economy in downstream operations
- Very high flow rates and large sample volume processing
- Excellent chemical stability
Their combination of high capacity, narrow specification range, high flow rate and low backpressure consistently reduces process cycle times and increases productivity. The Capto™ product range comprises modern media that meet the demands of large-scale biopharmaceutical manufacturing today.
Specifications
- Catalog No:
- 77509-278
- 77509-280
- pH Range:
- 3 - 13
- 3 - 13
- Storage:
- 4...30 °C, 20% Ethanol+
- 4...30 °C, 20% Ethanol+
Specifications
Frequently Bought Together