Order Entry
United States
Orders LinkContactUsLinkComponent
TE Buffer

TE Buffer

Supplier:   RPI
TE Buffer
Supplier:  RPI
Certificates
TE Buffer

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

TE buffer, at 1X concentration, is a commonly used solution, particularly in molecular biology and biochemistry. TE stands for Tris-EDTA, as it contains Tris (tris(hydroxymethyl)aminomethane) and EDTA (ethylenediaminetetraacetic acid).

  • It is also used in DNA and RNA quantification assays, such as spectrophotometry and fluorometry

TE buffer provides a stable environment for DNA and RNA by protecting them from degradation. Tris acts as a buffering agent, maintaining a stable pH, while EDTA chelates divalent metal ions, which are cofactors for nucleases, thus inhibiting their activity and protecting nucleic acids from enzymatic degradation.

TE buffer is commonly used in various molecular biology techniques, such as DNA and RNA extraction, storage of nucleic acids, DNA and RNA gel electrophoresis, and PCR (polymerase chain reaction) assays.

Caution: Research or further manufacturing use only, not for food or drug use.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77510-466
  • 77510-464
  • Appearance:
  • Colorless liquid
  • Colorless liquid
  • Storage Temperature:
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Concentration
    Size
    Application
    Availability
    Price
  • 77510-466
    Tris-EDTA
    Tris-EDTA
    1X
    500 ml
    Biological buffers
    Log in for availability
    Log in for availability
    Each
    Each - $48.62
    $48.62
    Each
       $48.62

    Supplier Number: T35150-500.0
    Specifications:
    More
    Specifications
    Cat. No.
    77510-466
    Appearance
    Colorless liquid
    Storage Temperature
    RT
  • 77510-464
    Tris-EDTA
    Tris-EDTA
    1X
    1 L
    Biological buffers
    Log in for availability
    Log in for availability
    Each
    Each - $77.82
    $77.82
    Each
       $77.82

    Supplier Number: T35150-1000.0
    Specifications:
    More
    Specifications
    Cat. No.
    77510-464
    Appearance
    Colorless liquid
    Storage Temperature
    RT

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.