ContactUsLinkComponent
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
  89002-206
undefined
KIMAX® Centrifuge Tube, Conical-Bottom, Graduated, with Pourout, Kimble Chase
  89002-206
 :  45186-50
  • Volume:
    50 ml
  • Cat. No.:
    89002-206
  • O.D.×L:
    29×118 mm
  • Supplier no.:
    45186-50
Tube has a short tapered bottom and a pour spout.
  • Graduated and calibrated to contain
Scale, legend and marking spot are durable white ceramic enamel. Graduated to 10 ml in 0.5 ml increments and from 10 to 50 ml in 1 ml increments.