ContactUsLinkComponent
Fluid Thioglycolate Medium (FTM), bioMerieux
Fluid Thioglycolate Medium (FTM), bioMerieux
Catalog # 29446-143
Supplier:  bioMerieux
CAS Number:  
Fluid Thioglycolate Medium (FTM), bioMerieux
Catalog # 29446-143
Supplier:  bioMerieux
Supplier Number:  302164
CAS Number:  
Specifications
  • Description:
    Fluid Thioglycollate Medium (FTM)
  • Cat. No.:
    29446-143
  • Volume:
    200 ml
  • Supplier no.:
    302164
Specifications
About this item
FTM is used for the cultivation of aerobic, anaerobic, and microaerophilic organisms.
  • Effective for sterility testing
FTM and FTM with polysorbate 80 are used as enrichment media for sterility testing, as well as detection of aerobic bacteria recommended by USP/EP/JP for sterility testing of pharmaceutical articles.