Reagent is provided as a 2mg/mL suspension in 20mM MES, 150mM NaCl, pH 6.2 buffer
Liposomal formulation optimized for maximal transfection efficiency of Sf9 insect cells. It features extremely low toxicity to the cells and can be used for both transient and stable transfection in both serum-free and serum-containing media. It can also be used for cotransfection of transfer plasmids with linearized virus DNA for the production of recombinant baculoviruses. Insect GeneJuice® is ideal for large-scale protein expression in suspension-culture transfection of Sf9 cells and when using pIEx™ and pBiEx™ vectors that contain the immediate early baculovirus promoter IE1. Just 1mL is sufficient for either 10 transfections in 10mL suspension culture flasks or 100 transfections in 35mm plates.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Bioproduction:Yes
- Cell Specific:Insect
- DNA:Yes