Orders LinkContactUsLinkComponent
Micro•Freeze™ Circuit Chiller, MicroCare
Micro•Freeze™ Circuit Chiller, MicroCare
Catalog #
Micro•Freeze™ Circuit Chiller, MicroCare
Catalog #
Supplier:  
Micro•Freeze™ Circuit Chiller, MicroCare
Micro•Freeze™ Circuit Chiller, MicroCare
Product Details & Documents
Chills to lower than –40°C in seconds
  • Micro-filtered to less than 0.2 microns
  • Uses nonflammable HFC-134a
  • Completely ozone- and plastic-safe and residue-free
Used to locate intermittent faults in delicate components and to provide heat sink protection when soldering.
Documents