Urea ACS, Ultrapure
Supplier: RPI
New Product
Certificates
About this item
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Boiling Pt:197
Molecular Weight:60.06 g/mol
Melting Pt:132.7
Merck Index:13,09935
Storage Temperature:Room Temperature
EINECS:200-315-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder or Pellets
- Assay (Titration, dried basis):99.0 - 100.5%
- Chlorides:≤5 ppm
- Heavy Metals (as Pb):≤0.001%
- Insoluble Matter:≤0.01%
- Melting Point:132 - 135 °C
- Residue on Ignition:≤0.01%
- Sulfate (SO4):Conforms
- BSE/TSE Free:BSE/TSE Free
- Grade:ACS
Specification Test Results
Frequently Bought Together