Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Supplier: RPI
Certificates
About this item
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Melting Pt:171...172 °C
Storage Temperature:Room Temperature
EINECS:201-064-4
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White Crystalline Powder
- Absorbance at 280 nm (1M Solution):≤0.05
- Absorbance at 300 nm (25% Solution):≤0.04
- APHA Color, 20% Aqueous solution:≤25
- Arsenic (As):≤5 ppm
- Assay (Titration, dried basis):99.5 - 101.0%
- Chloride:≤1 ppm
- Copper (Cu):≤1 ppm
- DNase, RNase, protease:Not Detected
- Electrophoresis:Pass Test
- Heavy Metals (as Pb):≤1 ppm
- Identification (IR):Pass
- Iron (Fe):≤1 ppm
- Magnesium (Mg):≤10 ppm
- Melting Point:168.0 - 172.0 °C
- Moisture (KF):≤0.5%
- pH (5% Solution):10.0 - 11.5
- Residue on Ignition:≤0.1%
- Solubility (40% Aqueous Solution):Clear
- Sulfate:≤5 ppm
- Turbidity (40% Solution):≤2.0
- UV Absorbance at 260 nm, 10% Aqueous solution:≤0.1
- UV Absorbance 280 nm (10% Solution in water):≤0.08
- UV Absorbance at 430 nm, 10% Aqueous solution:≤0.009
- BSE/TSE Free:BSE/TSE Free
- Non Animal Origin:Non Animal Origin
- Penicillin or other Beta Lactams:None Present
- Country of Origin:Report
Specification Test Results
Frequently Bought Together