Order Entry
United States
ContactUsLinkComponent
CellPro™ E-MEM, 1X, 1 L (without glutamine), Molecular Biology Grade, Sterile Filtered
CellPro™ E-MEM, 1X, 1 L (without glutamine), Molecular Biology Grade, Sterile Filtered
Catalog # 104028-066
Supplier:  Alkali Scientific
CAS Number:  
undefined
CellPro™ E-MEM, 1X, 1 L (without glutamine), Molecular Biology Grade, Sterile Filtered
Catalog # 104028-066
Supplier:  Alkali Scientific
Supplier Number:  EMEM001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    CellPro™ E-MEM, 1X, 1 L (without glutamine)
  • Size:
    1000 ml
  • Cat. No.:
    104028-066

Specifications

About this item

Achieve optimal cell growth with Alkali Scientific CellPro™ E-MEM, 1X, provided in a one liter volume without glutamine, molecular biology grade, and sterile filtered for reliability.

  • For molecular biology
  • Liquid medium
  • 0.2 μm filtered

CellPro™ Eagle's Minimum Essential Medium (E-MEM), offered in a convenient 1X concentration and 1 L volume without glutamine, serves as a premium choice for cell culture applications in molecular biology research. Formulated with meticulous attention to detail and meeting stringent quality standards, this sterile filtered medium ensures optimal conditions for cell growth and proliferation. Its exclusion of glutamine provides flexibility for researchers to tailor the medium to their specific experimental requirements. Packaged under sterile conditions to maintain integrity and purity, CellPro™ E-MEM offers researchers a reliable solution for a wide range of cell culture experiments, facilitating consistent and reproducible results in molecular biology studies.