Order Entry
United States
ContactUsLinkComponent
CellPro™ Synthetic Defined (Liquid), SD (Minimal) Media, Sterile Filtered
CellPro™ Synthetic Defined (Liquid), SD (Minimal) Media, Sterile Filtered
Catalog # 104027-550
Supplier:  Alkali Scientific
CAS Number:  
undefined
CellPro™ Synthetic Defined (Liquid), SD (Minimal) Media, Sterile Filtered
Catalog # 104027-550
Supplier:  Alkali Scientific
Supplier Number:  DP609
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Media Format:
    Liquid
  • Description:
    CellPro™ SD (minimal) media
  • Size:
    500 ml
  • Cat. No.:
    104027-550

Specifications

About this item

Provide defined growth conditions for yeast cultures with Alkali Scientific CellPro™ Synthetic Defined (Liquid) media, offered in a 500ml volume and sterile filtered for reliability.

  • For molecular biology
  • Liquid medium
  • 0.2 μm filtered

Synthetic Defined (Liquid) Media, meticulously formulated to molecular biology standards, provides researchers with a reliable solution for yeast culture applications. This sterile filtered, 500ml formulation offers optimal purity and sterility, ensuring consistent and reproducible results in molecular biology experiments. Suitable for a variety of yeast-based studies, Alkali Scientific Synthetic Defined (Liquid) Media facilitates robust growth and experimental success in the laboratory.