CellPro™ Tryptone, Molecular Biology Grade, Sterile Filtered
Provide essential amino acids for bacterial growth with Alkali Scientific CellPro™ Tryptone, meeting molecular biology grade standards and provided sterile filtered for purity.
- For microbiology
- Sterile
- Filtered
CellPro™ Tryptone is a high-quality peptone derived from casein, widely used as a nutrient source in microbial culture media. With its rich amino acid composition, this molecular biology-grade tryptone supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ Tryptone an essential component in various laboratory applications, including bacterial culture, recombinant protein production, and microbiological research.
Frequently Bought Together