ContactUsLinkComponent
Konform® SR Conformal Coating, ITW Chemtronics®
  47732-654
UL Listed
undefined
Konform® SR Conformal Coating, ITW Chemtronics®
  47732-654
 :  CTSR-1
UL Listed
  • Volume:
    3.8 L (1 gal.)
  • Cat. No.:
    47732-654
  • Supplier no.:
    CTSR-1
Formulated to provide maximum flexibility for extreme temperatures, this transparent coating protects both rigid and flexible printed circuit boards for the highest long-term protection in adverse operating conditions
UL listed.
Meets MIL-I-46058C, Type SR. Usable temperature range: –65° to 200°C (–85° to 390°F).