Sterile Shoe Covers, Class 10 (ISO 4)
Supplier: Alkali Scientific
Certificates
About this item
Step into sterile environments confidently with Alkali Scientific™ class 10 (ISO 4) white polypropylene sterile shoe covers.
- Non-woven spunbonded polypropylene
- Heavy-duty, slip-resistant sole
- Practical and durable 16" length
- Pre-sterilized for use in aseptic work environments
- Class 10 (ISO 4) compatible
These shoe covers offer a sterile solution for maintaining cleanliness in controlled environments. Constructed from white polypropylene material and meeting class 10 (ISO 4) standards, they provide protection against particulate matter and are suitable for use in sensitive settings such as cleanrooms and laboratories.
Specifications
- Catalog No:
- 104028-566
- 104028-796
- Fabric:
- Polypropylene
- Polypropylene
- Garment application:
- Cleanroom
- Cleanroom