About this item
Ensure sterile pipetting with Alkali Scientific™ PurePoint™ sterile 'V' filter pipet tips.
- Sterile
- 'V' design
- Integrated hydrophobic filter
- Universal compatibility
- Ergonomic design
PurePoint™ filter 'V' pipet tips are the pinnacle of laboratory precision, meticulously engineered to elevate pipetting accuracy and maintain sterile conditions throughout your liquid handling tasks. Crafted from premium-grade 100% polypropylene, these filter pipette tips are designed to optimize sample recovery and minimize residual liquid, reducing sample waste and enhancing pipetting precision. Each tip is equipped with an integrated hydrophobic filter, acting as a protective barrier against aerosols to prevent sample carryover and cross-contamination.
With wide compatibility across various pipette models, ergonomic design for comfortable handling, and a commitment to sterility and contamination control, PurePoint™ filter 'V' pipet tips are the trusted choice for researchers demanding excellence in their laboratory procedures.