Classic™ Sterile, Racked Pipet Tips, Universal Fit
Supplier: Alkali Scientific
Certificates
About this item
Experience consistent and reliable pipetting with Classic™ sterile, racked pipet tips, featuring a universal fit for various pipets.
- Fine point design
- Certified Rnase, Dnase and pyrogen free
- Universal Fit on research grade standard shaft pipets
- Ultra clear polypropylene, clear, yellow, blue
- Withstands organic solvents
- Low-retention
- Pre-Sterile, shrink-wrapped
- No need to autoclave
- each rack is Lot identified
Alkali Scientific™ Classic™ sterile racked pipette tips are designed to fit a wide variety of research grade pipets. Standardized fit and a stable rack make this the tip of choice for many multichannel users. Often, competitor tips won't align during multichanneling. Eliminate this frustration with this Classic™ series tip. Tips are standard 9 mm 'on center' to ensure a perfect fit. Semi-flexible walls and a series of subtle sealing rings help ensure a secure fit to improve overall pipetting accuracy. Seating tips and ejecting tips becomes an easier task with Alkali Scientific™ sterile racked pipette tips.
Frequently Bought Together