About this item
Minimize sample retention with Alkali Scientific™ Rainin LTS™ LiteTouch™ reload stackable tips - low retention, ensuring efficient pipetting in your laboratory.
- Stackable design reduces storage
- LTS, XTS Ranin®* compatible
- Environmentally friendly - 75% less waste
- Great for multichannels
- Low forces - reduces tip ejection force up to 85%
- Low retention - most precise, accurate delivery
- Great fit on Rainin®* LiteTouch™ pipets
- RNAse, DNAse, pyrogen free
- Sterile
Experience precision and sustainability with Rainin LTS™ and XLS™ LiteTouch™ reload stackable tips. Crafted for optimal liquid handling, these tips ensure the utmost accuracy with low retention design while reducing waste by up to 75% through environmentally friendly packaging. With compatibility with Rainin® LiteTouch™ pipets, they offer seamless integration and up to 85% less force for ejection, promoting ergonomic comfort during repetitive tasks. Moreover, they maintain sample integrity with RNAse, DNAse, and pyrogen free composition. Elevate your pipetting experience with Rainin LTS™ and XLS™ LiteTouch™ reload stackable tips, where precision meets sustainability.