Orders LinkContactUsLinkComponent
Gel Loading Tips, 200 µl
Gel Loading Tips, 200 µl
Marketsource
Supplier:   Alkali Scientific
Gel Loading Tips, 200 µl
Supplier:  Alkali Scientific
Marketsource
Certificates
Gel Loading Tips, 200 µl
Gel Loading Tips, 200 µl
3 Options Available Below
Product Details & Documents
Accurately load gels with Alkali Scientific™ gel loading tips, designed for precise delivery of 200 µl volumes.
  • Designed for precise and accurate sample loading into gels
  • Patented press-to-open racks provide easy, one-handed opening
  • Universal fit to accommodate most pipettor brands
  • Multichannel-friendly with a standard 96-tip arrangement
  • For use in pharmaceutical and food industry, and molecular biology
  • Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors and tested pyrogen-free
  • Color: -00 Natural
Ensure accurate and efficient sample loading with gel loading tips, each capable of holding up to 200 μl of sample. Designed specifically for use in gel electrophoresis applications, these tips provide precise control over the placement of your samples into gel wells. Their narrow, elongated shape allows for easy access to even the smallest wells, minimizing the risk of cross-contamination between samples. Crafted from high-quality materials, these tips offer excellent clarity, allowing you to monitor sample dispensing with ease. Whether you're loading DNA, RNA, or protein samples, gel loading tips provide the reliability and precision necessary for successful electrophoresis experiments.
Packaging: Packaged 96 tips/rack, 2 racks/unit, 5 units/case.
Product Family Options
Product Information
  • Volume
    Sterile
    Packed
    Availability
    Price
  • 0.1 - 200 µl
    +
    2 racks of 96 tips
    Log in for availability
    Log in for availability
    Each
    Each - $41.48
    $41.48
    Each
       $41.48
    Minimum order quantity is 1
    Supplier Number: RGL100
    Specifications:
    Marketsource
    Marketsource
  • 0.1 - 200 µl
    +
    2 racks of 96 tips, 5 packs/case
    Log in for availability
    Log in for availability
    Each
    Each - $191.06
    $191.06
    Each
       $191.06
    Minimum order quantity is 1
    Supplier Number: RGL102
    Specifications:
    Marketsource
    Marketsource
  • 0.1 - 200 µl
    +
    4×200 tips, 800/pack
    Log in for availability
    Log in for availability
    Each
    Each - $191.06
    $191.06
    Each
       $191.06
    Minimum order quantity is 1
    Supplier Number: RGL304
    Specifications:
    Marketsource
    Marketsource