Potassium Chloride Conductivity Standard, 12880 µS/cm (12.88 mS/cm) at 25 °C
RCR5888130-1A
:
- Pack type:1 L plastic
- Reference temperature:25 °C
- Cat. No.:RCR5888130-1A
- Conductivity:12880 µS/cm
- Certified traceable to NIST Standard Reference Materials
- Tested in ISO 17025 Accredited Laboratories
Calibrate with confidence using RICCA Conductivity Standards. Suitalbe for use with any probe. Tested to the tightest specifications in the industry, you can count on RICCA conductivity buffers to produce accurate and reproducible readings with every pour.