Order Entry
United States
ContactUsLinkComponent
Conductivity Standard, 200000 µS/cm (200 mS/cm) at 25 °C
Conductivity Standard, 200000 µS/cm (200 mS/cm) at 25 °C
Catalog # RCR2249200500A
Supplier:  Ricca Chemical
New Product
CAS Number:  
undefined
Conductivity Standard, 200000 µS/cm (200 mS/cm) at 25 °C
Catalog # RCR2249200500A
Supplier:  Ricca Chemical
Supplier Number:  R2249200-500A
New Product
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    500 ml plastic
  • Environmentally Preferable:
  • Reference temperature:
    25 °C
  • Cat. No.:
    RCR2249200500A
  • Conductivity:
    200000 µS/cm

Specifications

About this item

  • Certified traceable to NIST Standard Reference Materials
  • Tested in ISO 17025 Accredited Laboratories

Calibrate with confidence using RICCA Conductivity Standards. Suitalbe for use with any probe. Tested to the tightest specifications in the industry, you can count on RICCA conductivity buffers to produce accurate and reproducible readings with every pour.