Specifications
- Sterility:Sterile filtered
- Description:Charcoal dextran stripped, Fetal Bovine Serum (FBS), USDA approved
- Size:500 ml
- Environmentally Preferable:
- Cat. No.:104027-746
- Packaging:Bottle
Specifications
About this item
Maintain cell culture purity with Alkali Scientific™ Charcoal Dextran Stripped FBS, provided in a 500 ml volume for consistent and reliable cell culture experiments.
- Triple 0.1 µm sterile-filtered
- Mycoplasma-tested and virus-screened
- Ships on dry ice; Store frozen at −20 to −10 °C
CellPro™ Charcoal Dextran Stripped Fetal Bovine Serum (FBS) offers researchers a specialized serum formulation optimized for hormone and steroid receptor assays, cell differentiation studies and other sensitive cell culture applications. Through a unique process of charcoal and dextran treatment, this serum product effectively removes unwanted hormones and steroids while preserving essential growth factors and nutrients necessary for cell growth and viability. With its meticulously controlled formulation and sterile packaging, CellPro™ Charcoal Dextran Stripped FBS provides researchers with a reliable and consistent serum source for their experimental needs, ensuring robust and reproducible results in cell culture studies. Whether used for hormone receptor assays or other demanding applications, CellPro™ Charcoal Dextran Stripped FBS stands as a trusted choice for researchers seeking high-quality and reliable serum products for their cell culture experiments.