ContactUsLinkComponent
VENOJECT® Multi-Sample Luer Adapter, Terumo
VENOJECT® Multi-Sample Luer Adapter, Terumo
Catalog # TEMN2000
Supplier:  Terumo
VENOJECT® Multi-Sample Luer Adapter, Terumo
Catalog # TEMN2000
Supplier:  Terumo
Supplier Number:  MN*2000T
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    VENOJECT® Multi-Sample Luer Adapter
  • Cat. No.:
    TEMN2000
  • Supplier no.:
    MN%2000T
Specifications
About this item
Sterile, disposable adapter fits into a standard infusion set transforming it into a winged blood collection set
This allows for multiple draws into evacuated tubes. Accepts standard tube holders.