D-(+)-Mannose
About this item
Caution: Research or further manufacturing use only, not for food or drug use.
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Storage Temperature:Room Temperature
- EINECS:222-392-4
4 Options Available Below
Product Details & Documents
Caution: Research or further manufacturing use only, not for food or drug use.
- CAS Number:3458-28-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00064122
- Molecular Weight:180.16 g/mol
- Storage Temperature:Room Temperature
- EINECS:222-392-4
Specifications for Product Family
Specification Test Results
- Appearance:White powder/off white powder
- Assay:≥99.0%
- Heavy Metals:≤10 ppm
- Loss on Drying:≤0.5%
- Melting Point:124 - 136 °C
- Optical Rotation:+13.0 - +15.0°
- Yeast and Mold:≤1000 cfu/g
- BSE/TSE Free:BSE/TSE Free
- Country of Origin:Report
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: M18000-25.0
- Supplier Number: M18000-100.0
- Supplier Number: M18000-500.0
- Supplier Number: M18000-1000.0
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



