Order Entry
United States
ContactUsLinkComponent
Human Recombinant HLA-A*02:01&B2M&MAGE-A10 (MHC-HM459)
Human Recombinant HLA-A*02:01&B2M&MAGE-A10 (MHC-HM459)
Supplier:  Kactus Bio
Certificates
undefined
Human Recombinant HLA-A*02:01&B2M&MAGE-A10 (MHC-HM459)
Supplier:  Kactus Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    C-His-Avi
  • Storage Conditions:
    Valid for 12 months from date of receipt when stored at –80 °C. Recommend to aliquot the protein into smaller quantities for optimal storage. Please minimize freeze-thaw cycles.
  • Endotoxin Content:
    Less than 1 EU per μg by the LAL method.
  • Gene ID:
    A0A140T913(HLA-A*02:01)&P61769(B2M)&GLYDGMEHL
  • Endotoxin-free:
    Y
  • Carrier-Free:
    Y
  • Animal-Free:
    N
  • Protein Synonyms:
    HLA-A*02:01&B2M&MAGE-A10 (GLYDGMEHL) Monomer
  • Protein/Peptide Name:
    HLA-A*02:01&B2M&MAGE-A10
  • Purity:
    <95% as determined by Tris-Bis PAGE; <95% as determined by HPLC
  • Molecular Weight:
    The protein has a predicted MW of 50.44 kDa. Due to glycosylation, the protein migrates to 53 - 63 kDa based on Tris-Bis PAGE result.
  • Sequence:
    Gly25-Thr305(HLA-A*02:01), Ile21-Met119(B2M) and GLYDGMEHL peptide
  • Endotoxin Level:
    Less than 1 EU per μg by the LAL method.
  • Formulation:
    Supplied as 0.22 μm filtered solution in PBS (pH 7.4).
  • Shipping Temperature:
    Shipped with dry ice.
  • Tested Applications:
    SDS-Page, HPLC

Specifications

Frequently Bought Together