Human Recombinant HLA-A*02:01&B2M&MAGE-A4, Biotin (MHC-HM437B)
Supplier: KACTUSBIO INC. MS
Certificates
About this item
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
MAGE-A4 and MAGE-A8 are type I membes of the Melanoma associated antigen (MAGE) family. The MAGE family is a large, highly conserved group of proteins that share a common MAGE homology domain. Both MAGE-A4 and MAGE-A8 antigen-presenting peptides can be presented by HLA-A*02:01.
Certifications: ISO 13485.
Delivery: One to two days.
Packaging: Recyclable.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:C-His-Avi
- Storage Conditions:Valid for 12 months from date of receipt when stored at –80 ℃. Recommend to aliquot the protein into smaller quantities for optimal storage. Please minimize freeze-thaw cycles.
- Endotoxin Content:Less than 1 EU per μg by the LAL method.
- Gene ID:A0A140T913(HLA-A*02:01)&P61769(B2M)&KVLEHVVRV
- Endotoxin-free:Y
- Carrier-Free:Y
- Animal-Free:N
- Protein Synonyms:HLA-A*02:01&B2M&MAGE-A4 (KVLEHVVRV) Monomer
- Protein/Peptide Name:HLA-A*02:01&B2M&MAGE-A4
- Purity:<95% as determined by Tris-Bis PAGE; <95% as determined by HPLC
- Molecular Weight:The protein has a predicted MW of 50.50 kDa. Due to glycosylation, the protein migrates to 52 - 62 kDa based on Tris-Bis PAGE result.
- Sequence:Gly25-Thr305(HLA-A*02:01), Ile21-Met119(B2M) and KVLEHVVRV peptide
- Endotoxin Level:Less than 1 EU per μg by the LAL method.
- Formulation:Supplied as 0.22 μm filtered solution in PBS (pH 7.4).
- Shipping Temperature:Shipped with dry ice.
- Tested Applications:SDS-Page, HPLC
Specifications
Frequently Bought Together