Human Recombinant HLA-A*02:01&B2M&P53 R175H (MHC-HM415T)
Supplier: Kactus Bio
Certificates
About this item
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
p53 is a tumor suppressor protein. Under stressful conditions, p53 tightly regulates cell growth by promoting apoptosis and DNA repair. When p53 becomes mutated, it loses its function, resulting in abnormal cell proliferation and tumor progression. Depending on the p53 mutation, it has been shown to form aggregates leading to negative gain of function of the protein. p53 mutant associated aggregation has been observed in several cancer tissues and has been shown to promote tumor growth.
Certifications: ISO 13485
Delivery information: One to two days.
Packaging: Recyclable.
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:C-His-Avi
- Storage Conditions:–20 to –80 °C for 12 months as supplied from date of receipt. –80 °C for 3 - 6 months after reconstitution. 2 - 8 °C for 2 - 7 days after reconstitution. Recommend to aliquot the protein into smaller quantities for optimal storage. Please minimize freeze-thaw cycles.
- Endotoxin Content:Less than 1 EU per μg by the LAL method.
- Gene ID:A0A140T913(HLA-A*02:01)&P61769(B2M)&HMTEVVRHC
- Reconstitution Instructions:Centrifuge the tube before opening. Reconstituting to a concentration more than 100 μg/ml is recommended. Dissolve the lyophilized protein in distilled water.
- Endotoxin-free:Y
- Carrier-Free:Y
- Animal-Free:N
- Protein Synonyms:P53|HLA-A|BCC7|TRP53|HLA-A*02:01&B2M&P53 R175H (HMTEVVRHC) Tetramer|MHC|TP53|LFS1|Antigen NY-CO-13|FLJ92943
- Protein/Peptide Name:HLA-A*02:01&B2M&P53 R175H
- Purity:<95% as determined by Tris-Bis PAGE; <95% as determined by HPLC
- Molecular Weight:The protein has a predicted MW of 258 kDa. Due to glycosylation, the protein migrates to 260 - 265 kDa under Non reducing (N) condition based on Tris-Bis PAGE result.
- Sequence:Gly25-Thr305(HLA-A*02:01),Ile21-Met119(B2M) and HMTEVVRHC peptide
- Endotoxin Level:Less than 1 EU per μg by the LAL method.
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS (pH 7.4). Normally 8% trehalose is added as protectant before lyophilization.
- Shipping Temperature:Shipped at ambient temperature.
- Tested Applications:ELISA, SDS-Page, HPLC
Specifications
Frequently Bought Together