Human Recombinant HLA-A*02:01&B2M&GP100, Biotin
Supplier: KACTUSBIO INC. MS
Certificates
About this item
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
Glycoprotein 100, gp100 or Melanocyte protein PMEL is 661 amino acids long and is a type I transmembrane glycoprotein enriched in melanosomes, which are the melanin-producing organelles in melanocytes. This protein is involved in melanosome maturation. The Human HLA-A*0201 gp100 (YLEPGPVTA) complex Protein is a complex of HLA-A*0201 of the MHC Class I and YLEPGPVTA peptide of the gp100.
Certifications: ISO 13485
Delivery: One to two days.
Packaging: Recyclable.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together