Human Recombinant HLA-A*02:01&B2M&GP100 (MHC-HM402)
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
Glycoprotein 100, gp100 or Melanocyte protein PMEL is 661 amino acids long and is a type I transmembrane glycoprotein enriched in melanosomes, which are the melanin-producing organelles in melanocytes. This protein is involved in melanosome maturation. The Human HLA-A*0201 gp100 (YLEPGPVTA) complex Protein is a complex of HLA-A*0201 of the MHC Class I, B2M and YLEPGPVTA peptide of the gp100.
: ISO 13485.
: One to two days.
: Recyclable.
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:C-His-Avi
- Storage Conditions:–20 to –80 ℃ for 12 months as supplied from date of receipt. –80 ℃ for 3 - 6 months after reconstitution. 2 - 8 ℃ for 2 - 7 days after reconstitution. Recommend to aliquot the protein into smaller quantities for optimal storage. Please minimize freeze-thaw cycles.
- Endotoxin Content:Less than 1 EU per μg by the LAL method.
- Gene ID:A0A140T913(HLA-A*02:01)&P61769(B2M)&YLEPGPVTA
- Reconstitution Instructions:Centrifuge the tube before opening. Reconstituting to a concentration more than 100 μg/ml is recommended. Dissolve the lyophilized protein in distilled water.
- Endotoxin-free:Y
- Carrier-Free:Y
- Animal-Free:N
- Protein Synonyms:Glycoprotein 100|MHC I|HLA-A*02:01&B2M&GP100 (YLEPGPVTA) Monomer|HLA0201|gp100|ME20-M|P100
- Protein/Peptide Name:HLA-A*02:01&B2M&GP100
- Purity:<95% as determined by Tris-Bis PAGE; <95% as determined by HPLC
- Molecular Weight:The protein has a predicted MW of 50.4 kDa. Due to glycosylation, the protein migrates to 52 - 62 kDa based on Tris-Bis PAGE result.
- Sequence:Gly25-Thr305(HLA-A*02:01),Ile21-Met119(B2M) and YLEPGPVTA peptide
- Endotoxin Level:Less than 1 EU per μg by the LAL method.
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS (pH 7.4). Normally 8% trehalose is added as protectant before lyophilization.
- Shipping Temperature:Shipped at ambient temperature.
- Tested Applications:ELISA, SDS-Page, HPLC
Frequently Bought Together