Human Recombinant HLA-A*02:01&B2M&MAGE-A4 (MHC-HM401T)
Supplier: Kactus Bio
Certificates
About this item
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
Melanoma-associated antigen 4 is a protein that in humans is encoded by the MAGEA4 gene. The MAGE- A4 antigen is among the most commonly expressed cancer testis antigens. The Human HLA-A*0201 MAGE-A4 (GVYDGREHTV) complex Protein is a complex of HLA-A*0201 of the MHC Class I, B2M and GVYDGREHTV peptide of the MAGE-A4.
Certifications: ISO 13485
Delivery information: One to two days.
Packaging: Recyclable.
Specifications
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:C-His-Avi
- Storage Conditions:–20 to –80 °C for 24 months as supplied from date of receipt. –80 °C for 3 - 6 months after reconstitution. 2 - 8 °C for 2 - 7 days after reconstitution. Recommend to aliquot the protein into smaller quantities for optimal storage. Please minimize freeze-thaw cycles.
- Endotoxin Content:Less than 1 EU per μg by the LAL method.
- Gene ID:A0A140T913(HLA-A*02:01)&P61769(B2M)&GVYDGREHTV
- Reconstitution Instructions:Centrifuge the tube before opening. Reconstituting to a concentration more than 100 μg/ml is recommended. Dissolve the lyophilized protein in distilled water.
- Endotoxin-free:Y
- Carrier-Free:Y
- Animal-Free:N
- Protein Synonyms:MAGE4A|MHC I|HLA0201|HLA-A*02:01&B2M&MAGE-A4 (GVYDGREHTV) Tetramer|CT1.4|MAGE4B|member 4|MAGE-X2|MAGE-A4
- Protein/Peptide Name:HLA-A*02:01&B2M&MAGE-A4
- Purity:<95% as determined by Tris-Bis PAGE; <95% as determined by HPLC
- Molecular Weight:The protein has a predicted MW of 258 kDa. Due to glycosylation, the protein migrates to 260 - 265 kDa under Non reducing (N) condition based on Tris-Bis PAGE result.
- Sequence:Gly25-Thr305(HLA-A*02:01),Ile21-Met119(B2M) and GVYDGREHTV peptide
- Endotoxin Level:Less than 1 EU per μg by the LAL method.
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS (pH 7.4). Normally 8% trehalose is added as protectant before lyophilization.
- Shipping Temperature:Shipped at ambient temperature.
- Tested Applications:ELISA, SDS-Page, HPLC
Specifications
Frequently Bought Together