Human Recombinant HLA-G&B2M&Peptide (HLG-HM41CT)
Supplier: KACTUSBIO INC. MS
Certificates
About this item
Quality validated and reproducible proteins provided at very competitive pricing.
- Best quality
- Reproducible supply
- Functionally validated
- Lowest price
HLA-G is a molecule that was first known to confer protection to the fetus from destruction by the immune system of its mother, thus critically contributing to fetal-maternal tolerance. The first functional finding constituted the basis for HLA-G research and can be summarized as such: HLA-G, membrane-bound or soluble, strongly binds its inhibitory receptors on immune cells (NK, T, B, monocytes/dendritic cells), inhibits the functions of these effectors, and so induces immune inhibition.
Certifications: ISO 13485
Delivery: One to two days.
Packaging: Recyclable.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together