Order Entry
United States
ContactUsLinkComponent
Penicillin : Streptomycin solution, 10,000 I.U. Penicillin, 10,000 ug/ml Streptomycin
Penicillin : Streptomycin solution, 10,000 I.U. Penicillin, 10,000 ug/ml Streptomycin
Catalog # 104028-038
Supplier:  Alkali Scientific
CAS Number:  
undefined
Penicillin : Streptomycin solution, 10,000 I.U. Penicillin, 10,000 ug/ml Streptomycin
Catalog # 104028-038
Supplier:  Alkali Scientific
Supplier Number:  FB84
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 ml
  • Environmentally Preferable:
  • Cat. No.:
    104028-038

Specifications

About this item

 :
UN:3248
CAS Number:1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)
 :113-98-4