$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Sodium hydrogen carbonate
Sodium hydrogen carbonate solution 7.5%, CellPro™
Sodium hydrogen carbonate solution 7.5%, CellPro™
104028-310
:
Alkali Scientific
:
Sodium hydrogen carbonate solution 7.5%, CellPro™
104028-310
:
Alkali Scientific
:
DS0258
:
Pack type:
Plastic bottle
Size:
100 ml
Cat. No.:
104028-310
:
:
84.01 g/mol
:
3248
:
144-55-8
:
NaHCO₃
:
MFCD00003528