Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium chloride
Potassium chloride solution 3M, CellPro™
Potassium chloride solution MMMA, CellPro™
Potassium chloride solution MMMA, CellPro™
104028-084
:
Alkali Scientific
:
Potassium chloride solution MMMA, CellPro™
104028-084
:
Alkali Scientific
:
DS0193
:
Pack type:
Plastic bottle
Size:
100 ml
Environmentally Preferable:
Cat. No.:
104028-084
:
Potassiumchloride
:
:
74.55 g/mol
:
3248
:
7447-40-7
:
KCl
:
MFCD00011360