Potassium chloride solution 1M, CellPro™
Catalog # 104026-650
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:104026-650
Specifications
About this item
:
:74.55 g/mol
UN:3248
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360