Magnesium sulfate solution 0.01M, CellPro™
Catalog # 104028-758
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 ml
- Cat. No.:104028-758
Specifications
About this item
:
:120.37 g/mol
UN:3248
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110