Zeocin® solution 1 mg/ml, CellPro™
Catalog # 104028-904
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:10 ml
- Cat. No.:104028-904
Specifications
About this item
:
UN:3248
CAS Number:11031-11-1
MDL Number:MFCD28100499