Streptomycin solution 10 mg/ml, CellPro™
Plastic bottle, 10 ml
Catalog # 104028-992
Marketsource
Supplier: Alkali Scientific
Supplier #: DS0178
Streptomycin solution 10 mg/ml, CellPro™
Catalog # 104028-992
Supplier: Alkali Scientific
Marketsource
CAS Number: 3810-74-0
Jump to:
Product Details & Documents
CellPro™ Streptomycin Solution provides researchers with a superior antibiotic option for molecular biology applications. Crafted to meticulous standards of purity and sterility, this solution ensures reliable inhibition of bacterial growth in experimental procedures. With its sterile filtered formulation and concentration of 10mg/mL in a 10mL volume, CellPro™ Streptomycin Solution offers researchers confidence and peace of mind in their molecular biology endeavors.
- For molecular biology
- Liquid Medium
- 0.2 μm filtered
- CAS Number:3810-74-0
- Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
- MDL Number:MFCD00037023
- Molecular Weight:1457.4 g/mol
- UN:3248
Specifications
Specifications
- Pack type:Plastic bottle
- Size:10 ml
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



