Kanamycin solution 30 mg/ml, CellPro™
Catalog # 104027-126
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:10 ml
- Cat. No.:104027-126
Specifications
About this item
:
UN:3248
CAS Number:59-01-8
MDL Number:MFCD00070289