G-418 Sulfate solution 0.2 mg/ml, CellPro™
Catalog # 104028-286
Supplier: Alkali Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:10 ml
- Cat. No.:104028-286
Specifications
About this item
:
:692.72 g/mol
UN:3248
CAS Number:108321-42-2
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314